It is a device which offers powerful, efficient performance with years of durability. The sheer versatility of the device due to a wide range of rotor options make it extemely useful across a range of requiremnets. Moreover, the device can also be programmed easily to match the unique needs of the user with its user-friendly interface. Also the brushless DC motor present in the device is dust-free and guarantees efficient performance without the need for constant maintenance.
- Microprocessor controlled for efficient performance even under high load
- Intuitive interface allows for easy and quick setting of parameters
- Lid lock safety feature to avoid opening of lid when the rotor is running
- One touch toggle between RCF and RPM with just a touch of a button
- Short spin feature for ultra-fast centrifugation and pelleting
- Adjustable ramp and brake speed
订购须知 Rotors have to be ordered separately.