VWR®, Dry bath
供应商: VWR
VWR®
VWRA460-0360EA 16898.4 CNY
VWRA460-0360
VWR®, Dry bath
干浴器 数显型干浴器
This instrument offers exceptional performance with a maintenance-free long life.
  • Aluminium-made blocks, resistant to chemical spillage and temperature shocks
  • Two heating blocks with long timer range from 1 minute to 99 hours 59 minutes and infinite mode
  • Large and clear display shows critical parameters; All set values are readily visible without toggle and no shared display
  • Programmable features; user can set and save up to 99 user defined programs (protocols)
  • Corrosion-free stainless steel instrument top and heating wells
  • Wide tube capacity range with convenient lifting gear for cleaning and disinfecting
  • Rapid heating and uniform temperature across the block throughout the operation
  • Built in safe temperature protection
Designed to provide a superior user experience, the device provides uniform heating across the blocks to ensure reliable performance. Additionally, numerous user-friendly features like variable parameter setting, auto temperature safety cut and more. ensures that the work in the lab is made easier without expending much effort or time.
Certifications: Protection Class IP 21.
订购须知 Blocks must be ordered separately.
Delivery information: PT1000 probe and block lifter supplied as standard accessories. Comes thermometer well for accurate measurement of temperature.