VWR®, PCR Plates, Classic
供应商: VWR
VWR®
VWRA734-3326EA 2088 CNY
VWRA734-3326 VWRA734-3328 VWRA734-3327 VWRA734-3329
VWR®, PCR Plates, Classic
微孔板 PCR板
VWR® PCR Classic plasticwares are DNAse-, RNAse- and endotoxin-free for hassle free application outcome in PCR labs.
  • Ultrathin wall helps with superior heat transmission
  • Sterilised as per SAL 10⁻⁶ gamma irradiation standard
Plates are made of virgin PP and the manufacturing facility is compliant to 100K cleanroom as well as ISO standard.