VWR®, Mini Tube Rotator
供应商: VWR
VWR®
VWRA444-1187EA 3051.36 CNY
VWRA444-1187
VWR®, Mini Tube Rotator
振荡器 旋转/ 摇摆式振荡器
VWR® Mini Tube Rotator is equipped with a high-performance AC motor for quiet and efficient experiments along with a compact design.
  • Compact design for bench space efficiency
  • AC synchronous motor for noiseless operation
  • Can accommodate up to 10 Blood collection tubes
  • Gentle yet effective mixing with circular motion
  • 30 RPM fixed speed with ± 2 RPM speed accuracy
  • Continuous/Infinite operation modes
  • Robust construction, strong metal design
  • Maximum operational weight capacity of 1Kg
Certifications: IP Protection Class according to DIN EN 60529: IP 21
订购须知 Supplied without accessories.