Serological pipettes, Thermo Scientific™
供应商: Thermo Fisher Scientific
NUNC170356TEA 0 CNY
NUNC170353T NUNC170354T NUNC170357T NUNC170358T NUNC170355T NUNC170356T
Serological pipettes, Thermo Scientific™
移液管和移液器 血清移液管
Choose from a broad selection of Thermo Scientific™ serological pipettes for daily lab work.
  • Colour-coded pipettes and packaging for convenient volume selection
  • Bold clear graduations with easy-to-read scale markings
  • Ensures dispensing accuracy of ±2%
  • Manufactured with high quality virgin resins
  • Sterilized by gamma irradiation per ISO 11137-2
  • Non-pyrogenic, non-cytotoxic
  • DNase/RNase, BSE/TSE and human DNA free
These sterile serological pipettes are ideal for aseptic liquid transfers.
Packaging: Individually wrapped. Paper/plastic wrapping allowing for easy opening.