Pentair Hardness/Iron Reduction Double-Open-End Liquid Filter Cartridges
供应商: Pentair
MFLB29830-60EA 0 CNY
MFLB29830-60
Pentair Hardness/Iron Reduction Double-Open-End Liquid Filter Cartridges
过滤器 滤筒
Prevents scale and rust buildup—ideal for water processing.
  • Constructed with Hexametaphosphate crystals
  • FDA-compliant
Hexametaphosphate Crystal Cartridge prevents scale and rust buildup — use with water heaters, air conditioning equipment, and many other types of water processing equipment. Food-grade hexametaphosphate dissolves slowly in water to treat scale, corrosion, and iron problems at various flow rates and feed concentrations. All materials are FDA-compliant.
Water being treated should have no more than 257 mg/L hardness, with a pH range of 6.5 to 9.0. Maximum temperature is 100 °F (38 °C). Cartridge is made from polypropylene (PP) shell with a polyester pad prefilter and a PP post filter.