Micropump Cavity-Style Gear Pump Heads for NEMA Type 56 C-face Motors
Smooth pulseless flow provides precise, leak-free delivery of fluids.
- Compact size and easy to service
- Cavity-style heads are designed to handle higher inlet pressures.
Magnetically driven, precision-geared cavity pump heads provide smooth, accurate, pulseless fluid delivery. The magnetic drive and a single seal keep fluids securely inside and potential contaminants out. Chemical resistant materials handle a wide range of fluids.
- Environmentally Preferable:
- Catalog No:
- MFLB73007-02
- MFLB73007-00
- MFLB73007-11
- Body material:
- 316 stainless steel
- 316 stainless steel
- 316 stainless steel
- Differential pressure:
- 6,9 bar (100 psi)
- 4,1 bar (60 psi)
- 3,4 bar (50 psi)
- Duty cycle continuous:
- Yes
- Yes
- Yes
- Gear material:
- PEEK
- PEEK
- PEEK
- Inlet connection:
- 0,375 NPT(F)
- 0,375 NPT(F)
- 0,75 NPT(F)
- Internal bypass:
- No
- No
- No
- Max. temperature:
- 93 °C (200 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- Max. viscosity:
- 1500 cP
- 1500 cP
- 2500 cP
- Min. temperature:
- –29 °C (–20 °F)
- –29 °C (–20 °F)
- –46 °C (–50 °F)
- Min. viscosity:
- 0,2 cP
- 0,2 cP
- 0,2 cP
- Outlet connection:
- 0,375 NPT(F)
- 0,375 NPT(F)
- 0,75 NPT(F)
- Reversible:
- -
- -
- +
- Seal material:
- TEV
- TEV
- PTFE
- Suction lift, wet:
- 610 mm of H₂O (24" H₂O)
- 610 mm of H₂O (24" H₂O)
- 610 mm of H₂O (24" H₂O)
- Warranty:
- 1 year
- 1 year
- 1 year
Product Information
- Flow rateMaximum Flow RateMax. motor RPMPk
- NEMA 56C Cavity Style Pump Head; 100psi/SS/PEEK/TEV; 3,48 ml/revNEMA 56C Cavity Style Pump Head; 100psi/SS/PEEK/TEV; 3,48 ml/rev73007-023,48 ml/rev13,92 L/min (3,7 gal./min)4000 rpm3161 * 316 件¥0.00
Specifications
Environmentally PreferableBody material316 stainless steelDifferential pressure6,9 bar (100 psi)Duty cycle continuousYesGear materialPEEKInlet connection0,375 NPT(F)Internal bypassNoMax. temperature93 °C (200 °F)Max. viscosity1500 cPMin. temperature–29 °C (–20 °F)Min. viscosity0,2 cPOutlet connection0,375 NPT(F)Reversible-Seal materialTEVSuction lift, wet610 mm of H₂O (24" H₂O)Warranty1 year - NEMA 56C Cavity Style Pump Head; 60psi/SS/PEEK/TEV; 3,48 ml/revNEMA 56C Cavity Style Pump Head; 60psi/SS/PEEK/TEV; 3,48 ml/rev73007-003,48 ml/rev13,92 L/min (3,7 gal./min)4000 rpm11 * 1 件¥0.00
Specifications
Environmentally PreferableBody material316 stainless steelDifferential pressure4,1 bar (60 psi)Duty cycle continuousYesGear materialPEEKInlet connection0,375 NPT(F)Internal bypassNoMax. temperature93 °C (200 °F)Max. viscosity1500 cPMin. temperature–29 °C (–20 °F)Min. viscosity0,2 cPOutlet connection0,375 NPT(F)Reversible-Seal materialTEVSuction lift, wet610 mm of H₂O (24" H₂O)Warranty1 year - NEMA 56C Cavity Style Pump Head; 50psi/SS/PEEK/PTFE; 7,7 ml/revNEMA 56C Cavity Style Pump Head; 50psi/SS/PEEK/PTFE; 7,7 ml/rev73007-117,7 ml/rev26,57 L/min (7 gal./min)3450 rpm11 * 1 件¥0.00
Specifications
Environmentally PreferableBody material316 stainless steelDifferential pressure3,4 bar (50 psi)Duty cycle continuousYesGear materialPEEKInlet connection0,75 NPT(F)Internal bypassNoMax. temperature93 °C (200 °F)Max. viscosity2500 cPMin. temperature–46 °C (–50 °F)Min. viscosity0,2 cPOutlet connection0,75 NPT(F)Reversible+Seal materialPTFESuction lift, wet610 mm of H₂O (24" H₂O)Warranty1 year