Tricorn Filter Kits
供应商: GE HEALTHCARE
11-0012-53 11-0012-54
GEHE11-0012-53EA 0 CNY
GEHE11-0012-53 GEHE11-0012-54
Tricorn Filter Kits
过滤器 过滤盘
Filter kits for Tricorn 5 and 10 columns are available in three porosities.
Choose the standard 7 µm porosity for most applications. Use the medium 18 µm porosity for the purification of polyclonal IgG antibodies or mAbs in combination with resins such as MabSelect SuRe pcc or MabSelect PrismA. Use the coarse filter (>25 µm) to reduce the risk of clogging your capture resin when working with large volumes or repeated loadings of clarified samples.
Delivery information: All filter kits include top and bottom filters and ethylene propylene diene monomer (EPDM) O-rings, five of each.