This compact machine is a unique combination of high speed microcentrifuge and tunable mixer in one device. Ideal for low volume applications in PCR diagnostic laboratories, biochemical clinical laboratories, sample preparation prior to PCR reactions, gel electrophoresis, enzyme reactions and immunology.
- Intuitive interface specially designed to ease parameter adjustments
- Push and spin mode designed to ensure the rapid centrifugation by pressing and holding the start/stop button
- Automatic rotor recognition system
- Safety lock
订购须知 Supplied with high speed rotor (15 000 min⁻¹ / 15 000×g) for 12×0,2/2 ml test tubes with magnetic cover, MIX rotor (12 000 min⁻¹ / 10 000×g) for mixing and centrifugation of 12×0,2/2 ml test tubes, and rotor for two immuno strips (12 000 min⁻¹ / 10 000×g).