Micro burettes, bang pattern, lateral stopcock, Schellbach, BLAUBRAND®
供应商: Brand
BRND24597EA 2800 CNY
BRND24597
Micro burettes, bang pattern, lateral stopcock, Schellbach, BLAUBRAND®
Borosilicate glass 3.3, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • With base
With interchangeable needle-valve stopcock with PTFE spindle (PTFE key in the intermediate stopcock).
Length: 900 mm except for 2 ml: 660 mm.
Certifications: DIN EN ISO 385
订购须知 Supplied with a batch certificate. Burettes are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.