Cell culture chambers, Nunc™ Cell Factory™ Systems with Active Gassing
供应商: Thermo Fisher Scientific
Nunc™
NUNC173238EA 0 CNY
NUNC173238 NUNC173240
Cell culture chambers, Nunc™ Cell Factory™ Systems with Active Gassing
生物反应器
PS, sterile. For industrial scale production of, for example, vaccines, monoclonal antibodies, or pharmaceuticals. The patented gas-flow system secures a controlled atmosphere in the culture trays by equal distribution of user-specified gas mix actively pumped through the pre-mounted filter. Particularly oxygen demanding and pH sensitive cells may benefit from the controlled atmosphere.
  • Gas is equally distributed between trays, as well as in each individual tray
  • Nunclon™∆ surface treatment ensures excellent conditions for cell attachment and growth
  • Available in 4, 10 and 40 tray versions for easy scale up
  • Low contamination risk
  • Compatible with existing manual and automated handling equipment from Nunc™
Tray L×W: 335×205 mm
Accessories information: *Start-up kit includes: PC connector, airvent filter, white filter adapter cap, cover caps, tube clamp and silicone tubing.