Centrifugal devices, Jumbosep™
Convenient and reliable concentration, purification and diafiltration of 15 to 60 ml biological samples. Concentrate 60 ml sample volumes to 4 ml in 30 minutes.
- High recoveries, typically greater than 90%
- Built-in deadstop prevents spinning to dryness
Can be used with rotors for 250 ml tubes. Economical device as sample reservoir and filtrate receiver can be sanitised or autoclaved and reused.
- Catalog No:
- PALLFD010K65
- PALLFD003K65
- PALLFD100K65
- PALLFD000K65
- PALLFD030K65
- PALLFD300K65
- Filtration area:
- 15,2 cm²
- 15,2 cm²
- 15,2 cm²
- 15,2 cm²
- 15,2 cm²
- 15,2 cm²
- Max. RCF:
- 3000 ×g
- 3000 ×g
- 3000 ×g
- 3000 ×g
- 3000 ×g
- 3000 ×g
- Dead volume:
- 0,2 ml
- 0,2 ml
- 0,2 ml
- 0,2 ml
- 0,2 ml
- 0,2 ml
- Ø×H:
- 60×113 mm
- 60×113 mm
- 60×113 mm
- 60×113 mm
- 60×113 mm
- 60×113 mm
- Sample volume:
- Max. 60000 µl
- Max. 60000 µl
- Max. 60000 µl
- Max. 60000 µl
- Max. 60000 µl
- Max. 60000 µl
- Temperature:
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
- 0 - 40 °C
Starter kits
- MWCOColour codePk
- Starter kit
Accessories
Specifications
Environmentally PreferableFiltration area15,2 cm²Max. RCF3000 ×gDead volume0,2 mlØ×H60×113 mmSample volumeMax. 60000 µlTemperature0 - 40 °C - Starter kit
Accessories
Specifications
Environmentally PreferableFiltration area15,2 cm²Max. RCF3000 ×gDead volume0,2 mlØ×H60×113 mmSample volumeMax. 60000 µlTemperature0 - 40 °C - Starter kit
Accessories
Specifications
Environmentally PreferableFiltration area15,2 cm²Max. RCF3000 ×gDead volume0,2 mlØ×H60×113 mmSample volumeMax. 60000 µlTemperature0 - 40 °C - Starter kit
Accessories
Specifications
Environmentally PreferableFiltration area15,2 cm²Max. RCF3000 ×gDead volume0,2 mlØ×H60×113 mmSample volumeMax. 60000 µlTemperature0 - 40 °C - Starter kit
Accessories
Specifications
Environmentally PreferableFiltration area15,2 cm²Max. RCF3000 ×gDead volume0,2 mlØ×H60×113 mmSample volumeMax. 60000 µlTemperature0 - 40 °C - Generic starter kit, no membrane insertsGeneric starter kit, no membrane insertsPALLFD000K65FD000K65—41 * 4 件¥0.00
Accessories
Specifications
Environmentally PreferableFiltration area15,2 cm²Max. RCF3000 ×gDead volume0,2 mlØ×H60×113 mmSample volumeMax. 60000 µlTemperature0 - 40 °C
Membrane inserts
- SeqDescriptionMWCOPk