Membrane filters, Whatman™
供应商: Whatman products (Cytiva)
Whatman™
WHAT7141-154EA 0 CNY
WHAT7141-154 WHAT10401621 WHAT10401706 WHAT10400806 WHAT10406814 WHAT7141-004 WHAT10409772 WHAT10401631 WHAT10400906 WHAT10401670 WHAT10401606 WHAT7153-104 WHAT10400714 WHAT10406871 WHAT10401714 WHAT10401726 WHAT10406970 WHAT10401664 WHAT10407970 WHAT10400909 WHAT10409770 WHAT7141-124 WHAT10401614 WHAT10401712 WHAT10400706 WHAT10401772 WHAT10400812 WHAT10400814 WHAT7141-104 WHAT10401612 WHAT10406572 WHAT7141-114 WHAT10400921 WHAT10401514 WHAT7141-204 WHAT7140-104 WHAT10408472 WHAT10400914 WHAT10400912 WHAT10405672 WHAT10401731 WHAT10401618 WHAT10409870 WHAT10401506 WHAT10401512 WHAT10400821 WHAT7148-002 WHAT10409714 WHAT10401770 WHAT10401672 WHAT7187-114 WHAT10400712 WHAT10409771 WHAT10409414 WHAT10409970 WHAT10408970 WHAT10406972 WHAT10409814 WHAT10400772 WHAT10406512 WHAT10406812 WHAT10409872 WHAT10401626 WHAT10401721
Membrane filters, Whatman™
过滤器 膜过滤器
Hydrophillic mixed cellulose ester membranes are composed of cellulose acetate and cellulose nitrate. These membranes are characterised by a smoother and more uniform surface than pure nitrocellulose filters. Also, the colour contrast provided by the filter surface facilitates particle detection and minimises eye fatigue. The ME range has a lower cellulose acetate content compared to the WME range of membranes.
  • Excellent contrast for easier particle detection
  • Grids are non toxic and do not inhibit bacterial growth, ensuring sample integrity
  • The membrane offers a high degree of internal surface area for greater adsorption of product
  • Higher dirt loading capacity
  • Biologically inert with good thermal stability
  • No surfactants to contaminate samples
  • Uniform microporous structure of membrane gives high flow rates
Gridded filters have clearly defined grid lines spaced at 3,1 mm or 5 mm intervals.