Membrane filters, MicroPlus and ME, Whatman™
供应商: Whatman products (Cytiva)
Whatman™
WHAT10407134EA 0 CNY
WHAT10407134 WHAT10407372 WHAT10408714 WHAT10407370 WHAT10407170 WHAT10407314 WHAT10407112 WHAT10407332 WHAT10407172 WHAT10406870 WHAT10407615 WHAT10409834 WHAT10407714 WHAT10407342 WHAT10409472 WHAT10407713 WHAT10407324 WHAT10409470 WHAT10407114 WHAT10408915 WHAT10407334 WHAT10407132 WHAT10407312 WHAT10407734 WHAT10408712 WHAT10406872
Membrane filters, MicroPlus and ME, Whatman™
过滤器 膜过滤器
Developed for microbiological quality control of food and beverages, pharmaceuticals and cosmetics. All membrane filters have a high contrast grid (3,1 mm) and are sterile, individually packed. The STL versions are compatible with Membrane-Butler and other membrane filter dispensers.
Developed for microbiological quality control of foods and beverages, pharmaceuticals and cosmetics.
MicroPlus cellulose nitrate membranes: Designed specifically for microbiological QC. These filters have high mechanical strength and flow rate. They are tested for bubble point, bacterial retention, flow rate, recovery rate and sterility.
  • Sterile, individually packed
  • All membrane filters have a high contrast grid (3,1 mm)
  • Type STL in dispenser boxes with 100 numbered membrane filters for easy removal and safe handling with the Membrane-Butler
ME mixed cellulose ester, hydrophilic membranes: These filters have excellent contrast for easy particle detection, high internal surface area for sample adsorption, high dirt loading capacity. They are biologically inert with good thermal stability. The uniform microporous structure delivers high flow rates.
ME membranes