Filter funnels, MicroFunnel™
供应商: Pall Laboratory
PALL4805EA 0 CNY
PALL4805 PALL4806 PALL4817 PALL4828 PALL4803 PALL4804 PALL4815 PALL4818 PALL4852 PALL4801 PALL4810 PALL4800
Filter funnels, MicroFunnel™
漏斗 膜过滤漏斗
Ready to use disposable filter funnels with 47 mm diameter filtration membrane for microbiological analysis. Eliminates the cleaning and sterilisation required with reusable funnels. Membrane easily removed for culturing on agar or broth medium.
Filtration area: 13,46 cm²
Ø×H:
100 ml: 61×76 mm with cover
300 ml: 88×91 mm with cover
Membranes:
GN-6 Metricel®: Mixed cellulose esters, white
Supor®: Polyethersulfone, white
Metricel® Black: Modified polyethersulfone, black
* with Petri dish lid kit
Ø×H: 100 ml: 61×76 mm with cover; 300 ml: 88×91 mm with cover
Packaging: Sterile, individually bagged.
* with Petri dish lid kit; ** bulk packed