Sedimentation cones, Imhoff type
供应商: Brand
BRND387462EA 1825 CNY
BRND387462 BRND387662
Sedimentation cones, Imhoff type
Borosilicate 3.3 glass
Graduation and inscriptions in highly contrasting white enamel.
Certifications: DIN 12672