Dry box gloves, butyl
供应商: North by Honeywell
NORH8B1532/9QEA 0 CNY
NORH8B1532/9Q
Dry box gloves, butyl
手套
Butyl provides the highest permeation resistance to vapors and gases, as well as exceptional resistance to a broad range of toxic chemicals, making it ideal for many critical applications. Also suitable for use in food processing.
  • Flexible and sensitive, even at low temperatures
  • Particularly suited to applications involving highly toxic chemical hazards, such as chemical weapons
  • Highly resistant to ketones (MEK, MIBK, acetone) and esters (tricresyl phosphate, amyl acetate, ethyl acetate)
  • Solvent based, multi-dip manufacturing process provides multiple layers of polymer (like a glove within a glove) which results in a glove of superior quality