Disposable half masks against gases, vapours and particulates, Freedom
供应商: Honeywell Safety Products
DALL1001598EA 0 CNY
DALL1001598 DALL1001591
Disposable half masks against gases, vapours and particulates, Freedom
Half face masks with integrated filters for occasional or frequent exposures to gases, vapours and particulates.
  • Quick and easy adjustment
  • Excellent fit and comfort due to a soft seal flange
  • Disposable - no maintenance, better hygiene and no filter handling
  • Expiration valve for easier breathing
Weight: 265 g
Certifications: EN 405, EN 149: 2001
Caution: *One size fits all