Serological pipettes, Falcon®
供应商: Corning
Corning® Falcon®
BDAA357550_COS BDAA357525_COS BDAA357543_COS
Serological pipettes, Falcon®
移液管和移液器 血清移液管
PS, transparent, graduated, sterile.
  • Coloured top with cotton plug and coloured packaging for easy identification
  • Negative graduations for extra capacity and reverse graduations (except 1 ml)
  • Non pyrogenic
Packaging: Individually wrapped in paper-peelable bags or bulk packed in bags.