Dispenser tips, PD-Tips
供应商: Brand
PD-Tips
BRND702438EA 1500 CNY
BRND702438 BRND702390 BRND702368 BRND702396 BRND702684 BRND702380 BRND702392 BRND702382 BRND702394 BRND702372
Dispenser tips, PD-Tips
吸头
These dispenser tips are made from high-quality materials (cylinder: PP/piston: HDPE, 0,1 ml: LCP).
  • Direct displacement principle: Particularly suitable for dispensing of fluids with high viscosity, high vapor pressure, and more
  • Sterile BIO-CERT® PD-Tips are free from DNA, RNase, ATP and endotoxins
PD-Tips fit with: HandyStep® electronic, Gilson® Repetman®, Rainin AutoRep™ E (automatic tip recognition); HandyStep®, Rainin AutoRep™ M, Eppendorf® Multipette® 4780 and EDOS® 5221.
Certifications: ISO 8655
订购须知 25 and 50 ml tips supplied with adapter. Available either non sterile or beta-sterile/endotoxin-free (individually wrapped), or in BIO-CERT® quality. Supplied with batch certificate.
Caution: * Set (20×0,5 ml/20×1 ml/ 20×1,25 ml/20×2,5 ml/20×5 ml/20×10 ml/20×12,5 ml)