Autoclave tapes
供应商: PDC Healthcare - TimeMed
TIMETSI-512-2EA 0 CNY
TIMETSI-501-2 TIMETSI-534-2 TIMETSI-512-2
Autoclave tapes
灭菌指示带
White adhesive crepe tape, for use in standard autoclaves.
  • Dark-coloured diagonal lines appear after 15 minutes of exposure in a steam autoclave set at +121 °C
  • Clean removal from the autoclaved object
  • Latex free
Autoclaved after 15 minutes of exposure at +121 °C.
Clean removal from the autoclaved object.