Transfer membranes, BioTrace™ NT
供应商: Cytiva (Formerly Pall Lab)
BioTrace™
PALL66485EA 1575 CNY
PALL66485 PALL66489
Transfer membranes, BioTrace™ NT
转移膜
Pure unsupported nitrocellulose membrane ideally suited for colony and plaque lifts and protein transfers.
  • Strong and durable, less likely to tear or crack
  • High binding capacity for proteins and nucleic acids
  • Low protein burn-through in electrophoretic transfers
Pore size: 0,2 µm
Typical thickness: 145 µm
Protein binding capacity: 209 µg/cm²