Digital cameras, VisiCam®
VWRI630-0778EA 6596 CNY
VWRI630-0778
Digital cameras, VisiCam®
Each VWR VisiCam™ utilises a high speed USB 2.0 interface to quickly capture high quality images with good colour and resolution.
  • Easy to use
  • Low energy consumption - powered via USB 2.0 connection
  • Capturing formats: jpeg, .bmp, .raw, .avi, .mpeg
  • System requirements - Windows2000, WindowsXP
The VisiCam™ analyser software provides most basic functions for image analysis such as marking, counting, measuring, storage, white balance and objective calibration.
Delivery information: Includes: 1× C-mount adapter for VWR microscopes only (for fitting onto other microscopes please contact your local VWR sales office), eyepiece adapter (0,5×), calibration slide and VisiCam™ analyser software.