Order Entry
Automatic burettes, Pellet pattern, with intermediate stopcock, Schellbach stripe, without reservoir, BLAUBRAND®
 :  BRAND
undefined
Automatic burettes, Pellet pattern, with intermediate stopcock, Schellbach stripe, without reservoir, BLAUBRAND®
 :  BRAND
Needle-valve stopcock (PTFE key in intermediate stopcock)
  •  
    Capacity
    Division
    Tolerance
    Pk
  • BRND22761
    22761
    10 ml
    0,02 ml
    0,02 ± ml
    1
    1 * 1 件
    1 * 1 件 - ¥0.00
    ¥0.00
    1 * 1 件
       ¥0.00
    Accessories
    Specifications
    Environmentally Preferable
  • BRND22762
    22762
    25 ml
    0,05 ml
    0,03 ± ml
    1
    1 * 1 件
    1 * 1 件 - ¥0.00
    ¥0.00
    1 * 1 件
       ¥0.00
    Accessories
    Specifications
    Environmentally Preferable
  • BRND22763
    22763
    50 ml
    0,1 ml
    0,05 ± ml
    1
    1 * 1 件
    1 * 1 件 - ¥0.00
    ¥0.00
    1 * 1 件
       ¥0.00
    Accessories
    Specifications
    Environmentally Preferable
Accessories
  •  
    Capacity
    Description
    Pk
  • 2000 ml (2 L)
    Reservoir bottle, clear glass
    1
    1 * 1 件
    1 * 1 件 - ¥0.00
    ¥0.00
  •  
    Seq
    Description
    Pk
    Price
  • 05
    Ball set with net, black, Size 5, (102 ml), natural rubber/PVC
    1
    35,10
    1 * 1 件
    1 * 1 件 - ¥0.00
    ¥0.00