Membrane filters, polycarbonate, black, Whatman™ Cyclopore™
供应商: Whatman products (Cytiva)
Whatman™ Cyclopore™
WHAT7063-2502EA 0 CNY
WHAT7063-2502 WHAT7063-4702 WHAT7063-2504
Membrane filters, polycarbonate, black, Whatman™ Cyclopore™
过滤器 膜过滤器
The Cyclopore black membrane filter is a polycarbonate track etch membrane (PCTE membrane) that provides a contrast background for epifluorescence and other microscopy applications.
  • High sample visibility, provided by the black membrane filter surface
  • Negligible absorption and adsorption of filtrate; nonhygroscopic
  • Temperature resistant up to 140 °C
  • Low tare weight
Cytiva's business offers a variety of Whatman PCTE membrane filters for gravimetric analyses.
Because of their low background fluorescence, microfiltration membranes with a dark surface can improve test sensitivity in epifluorescence. Cyclopore black membrane filters offer a consistent contrasting background for excellent visibility during particle analysis.
Browse the range of Whatman track-etched membranes to find other Cyclopore membranes and Nuclepore application-specific membranes for organic halide adsorption and electron microscopy.