反式-2-癸烯醛 95%

供应商: Thermo Scientific
Warning

ACRO298190250EA 0 CNY
ACRO298190250
反式-2-癸烯醛 95%
反式-2-癸烯醛
分子式: C₁₀H₁₈O
分子量: 154,25 g/mol
沸点: 78…80 °C (3 mmHg)
闪点: 96 °C
存储条件: Refrigerator
MDL编号: MFCD00014679
CAS号: 3913-81-3
EINECS: 223-474-2
UN号: 3082
ADR: 9,III

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4600 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR