Bioassay dish, treated
供应商: Corning
Corning®
CORN431110EA 1913 CNY
CORN431110
Bioassay dish, treated
培养皿 生物活性皿
Optically clear PS. Treated for optimal cell attachment.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilised by gamma radiation
  • Certified non-pyrogenic