3-氯苯肼 97%
供应商: Thermo Scientific
|
Warning
|
ACRO167361000EA
0
CNY
ACRO167361000
ACRO167360250
3-氯苯肼 97%
3-氯苯肼
分子式:
C₆H₈Cl₂N₂ 分子量: 179,05 g/mol 溶点: 242 °C (dec.) |
MDL编号:
MFCD02656655 CAS号: 2312-23-4 EINECS: 219-005-6 |
产品技术规格
Appearance | White to light beige to pink Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...