Bioreactor chambers, Cellbag™
供应商: Cytiva
Cellbag™
GEHECB0002L10-03EA 0 CNY
GEHECB0002L10-03
Bioreactor chambers, Cellbag™
生物反应器
Presterile, single-use chambers for the non-invasive mixing of fluids using a WAVE Bioreactor™.
  • Designed to provide high mechanical strength and bio-inert fluid contact
  • Fluid contact layer is a medical grade low density polyethylene
  • Outer non-contact layer is made of low density polyethylene, EVA or nylon/EVOH copolymers
Other sizes available. Please enquire for further details.