2-羟基-5-甲氧基苯甲酸 98%

供应商: Thermo Scientific

Synonyms: 5-甲氧基水杨酸

ACRO163870100EA 0 CNY
ACRO163870100
2-羟基-5-甲氧基苯甲酸 98%
2-羟基-5-甲氧基苯甲酸
分子式: C₈H₈O₄
分子量: 168,15 g/mol
沸点: 327 °C (1013 hPa)
溶点: 142…146 °C
存储条件: Ambient
MDL编号: MFCD00002459
CAS号: 2612-02-4
EINECS: 220-037-8

现在订购

产品技术规格

Appearance Beige Crystalline powder
Infrared spectrum Conforms
Melting point 142 to 146 °C
HPLC ≥97.5 %
Residue after ign. (sulfated) ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR