氯化钾

供应商: APPLICHEM LIFESCIENCE

APLIA2939.0500EA 0 CNY
APLIA2939.0500
氯化钾
氯化钾
分子式: KCl
分子量: 74,55 g/mol
沸点: 1420 °C (1013 hPa)
溶点: 773 °C
密度: 1,98 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00011360
CAS号: 7447-40-7
EINECS: 231-211-8
Merck Index: 13,07704

现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR