Tubing connectors, Y-shaped, SP Bel-Art
供应商: Bel-Art
SCIENCEWARE®
BELAF196120000EA 604.19 CNY
BELAF196120000 BELAF196130000 BELAF196150000 BELAF196140000 BELAF196160000 BELAF196180000
Tubing connectors, Y-shaped, SP Bel-Art
连接器 管接头
Y-shaped connectors for tubing, made of chemical-resistant polypropylene, for use with a wide range of liquids or gases.
  • Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable