Culture media, ready to use, in ampoules, Whatman™
供应商: Cytiva
WHAT10496106EA 0 CNY
WHAT10496106 WHAT10496101 WHAT10496102 WHAT10496103 WHAT10496104
Culture media, ready to use, in ampoules, Whatman™
Nutrient media ( 2 ml ampoules) specifically suited to the cultivation of colonies after microfiltration.
  • Optimal media stability, sterility and reproducibility
  • Manufacturing process and product checks according to requirements of pharmacopoeia
  • Batch-specific quality certificate in each pack
Ready to use media can considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross-contamination. The wide range of media available satisfies even the most specialised customer requirements.
  • Optimal media stability, sterility and reproducibility
  • Manufacturing process and product checks according to requirements of pharmacopoeia
  • Batch-specific quality certificate in each pack
Ready to use media can considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross-contamination. The wide range of media available satisfies even the most specialised customer requirements.