Benzonase® ELISA kit II

供应商: Merck

Benzonase®
MERP1.01681.0001EA 0 CNY
MERP1.01681.0001
Benzonase® ELISA kit II
检测产品 ELISA试剂盒
For the immunological detection of Benzonase®.

Kit comprises:
Five microtitre plates: Each plate contains 8 single strips (12 wells/strip) coated with antibody specific to Benzonase®
Reagent A (0,6 ml): Benzonase® standard solution (purity grade I, 99%); concentration 5 µg/ml in buffer solution
Reagent B (0,6 ml): POD conjugated rabbit antibodies against Benzonase® in stabilising buffer solution
Reagent C (100 ml): TMB Substrate
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR