Centrifuge tubes and racks, CUBE 2UBE®, SP Bel-Art
供应商: Bel-Art
SCIENCEWARE®
BELAF378530001EA 0 CNY
BELAF378530001 BELAF378560000 BELAF378511000
Centrifuge tubes and racks, CUBE 2UBE®, SP Bel-Art
管子 微管
  • Can be centrifuged up to 14 000×g
  • Polypropylene; can be frozen to −20 °C
  • Compatible with Beckman BioMek 1000, Tecan, Hamilton and other robotic workstations
  • Each tube has a capacity of 2,2 or 211 ml per rack of 96 tubes
Offers the advantages and convenience of pipetting 2,2 ml per tube in a 8×12 grid, 96-well format.