TRIS-glycine-SDS buffer, READY-PACK™, Ultra Pure Grade
供应商: VWR Life Science
VWRC0147-40LEA 2110 CNY
VWRC0147-40L
TRIS-glycine-SDS buffer, READY-PACK™, Ultra Pure Grade
TG-SDS buffer for preparation of Laemmli SDS-PAGE running buffer.
The 40 litre pack size includes sufficient powder to prepare 40 litres of a 1X solution (0,025 M Tris, 0,192 M glycine and 0,1% SDS). Each Ready-Pack™ prepares 1 litre of a 10X solution.