Spatulas, micro
供应商: Corning Life Sciences
Corning®
CORN3012EA 1300 CNY
CORN3012 CORN3013
Spatulas, micro
样品勺
PS, white. These micro spatulas feature a V-shaped scoop (capacity < 0,5 ml) at one end and either a flat tapered or a flat rounded blade at the other end.
  • Free from RNase and DNase, non pyrogenic
  • Anti-static and sterile
  • Each micro spatula is individually packaged