Temperature probes
供应商: Cowie
COWIPC707.608.2001EA 890 CNY
COWIPC707.608.2001 COWIPC707.512.2001 COWIPC707.612.1501 COWIPC707.612.2501 COWIPC707.608.2501 COWIPC707.512.2501 COWIPC707.512.1501 COWIPC707.612.2001 COWIPC707.516.1501 COWIPC707.516.2501 COWIPC707.616.2501 COWIPC707.616.1501 COWIPC707.508.2501 COWIPC707.608.1501 COWIPC707.508.1501 COWIPC707.516.2001 COWIPC707.616.2001 COWIPC707.508.2001
Temperature probes
These baffle probes are suitable for reaction systems to increase agitation and promote mixing whilst also acting as temperature probe. PTFE encapsulated to ensure purity and chemical resistance and have a stainless steel core for rigidity.
  • Features 1 m long PFA insulated cable
Delivery information: Platinum probes supplied without plug and thermocouple probes supplied with miniplug not fitted.