4-甲氧基苯甲酰胺 ≥98.0% (通过GC)

供应商: TCI EUROPE
Warning

Synonyms: 对茴香酰胺

TCIAM1679-5GEA 240 CNY
TCIAM1679-5G
4-甲氧基苯甲酰胺 ≥98.0% (通过GC)
4-甲氧基苯甲酰胺
分子式: C₈H₉NO₂
分子量: 151,16 g/mol
沸点: 295°C
溶点: 130 °C
MDL编号: MFCD00007995
CAS号: 3424-93-9
EINECS: 222-319-6
https://cn.vwr.com/stibo/web/std.lang.all/59/76/28975976.jpg

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 164.0 to 168.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR